Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10519689 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519645 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519666 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519651 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519665 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519681 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519683 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519649 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519662 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519664 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519682 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519648 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519674 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10528155 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519672 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519653 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519654 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519682 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519690 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519666 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519650 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519649 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519680 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10528156 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519680 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519674 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10528155 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519678 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519673 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519673 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519664 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519648 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519646 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519665 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519672 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519690 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519650 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519689 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519653 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519681 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519651 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519654 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519644 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519661 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519688 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519677 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519652 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10528154 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519688 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10528154 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519671 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519644 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519671 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519652 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519661 |
- |
- |
Mouse Genome 430 2.0 Array |
1452794_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1452794_x_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0500000653.mm.1 |
TC0500000072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000653.mm.1 |
NR_001586.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000673.mm.1 |
TC0500000075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000673.mm.1 |
NM_001037919.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000673.mm.1 |
NM_001037919.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000698.mm.1 |
TC0500000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000698.mm.1 |
NM_001177510.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000698.mm.1 |
NM_001177510.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000699.mm.1 |
TC0500000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000699.mm.1 |
NM_001177510.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000699.mm.1 |
NM_001177510.1 |
Refseq |
Mouse Expression Set 430 |
1452794_x_at |
- |
- |
Mouse Expression Set 430 |
1452794_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1452794_x_at |
- |
- |
Mouse Expression Set 430 |
1452794_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1452794_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1452794_x_at |
- |
- |