Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039185.1 | NP_001034274.1 | 6 - 7 | 1447 | 111 | |
NM_011926.2 | NP_036056.2 | 4 - 5 | 907 | 111 | ||
GenBank mRNA | AB236328.2 | - | 4 - 5 | 816 | 111 | |
AB236330.2 | - | 5 - 6 | 1101 | 111 | ||
X67279.1 | - | 6 - 7 | 1459 | 111 | ||
X67281.1 | - | 6 - 7 | 1376 | 111 | ||
X67282.1 | - | 4 - 5 | 919 | 111 | ||
X67283.1 | - | 4 - 5 | 836 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |