Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077361.1 | NP_001070829.1 | 3 - 4 | 445 | 96 | |
NM_001077362.2 | NP_001070830.1 | 2 - 3 | 288 | 96 | ||
NM_001287800.1 | NP_001274729.1 | 2 - 3 | 232 | 96 | ||
NM_010211.3 | NP_034341.2 | 3 - 4 | 341 | 96 | ||
GenBank mRNA | AF114380.1 | - | 1 - 2 | 202 | 96 | |
AK128904.1 | - | 3 - 4 | 315 | 96 | ||
AK158966.1 | - | 3 - 4 | 373 | 96 | ||
BC029024.1 | - | 2 - 3 | 233 | 5012394 | 96 | |
BC031120.1 | - | 1 - 2 | 182 | 4989977 | 96 | |
BC059009.1 | - | 1 - 2 | 167 | 96 | ||
U41739.1 | - | 2 - 3 | 232 | 96 | ||
U77039.1 | - | 3 - 4 | 285 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |