Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081134.1 | NP_001074603.1 | 1 - 2 | 861 | 111 | |
GenBank mRNA | BC147251.1 | - | 1 - 2 | 793 | 9056302 | 111 |
BC147252.1 | - | 1 - 2 | 793 | 9056297 | 111 | |
BC147715.1 | - | 1 - 2 | 793 | 9087474 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |