Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081680.1 | NP_001075149.1 | 2 - 3 | 230 | 77 | |
GenBank mRNA | BC118036.1 | - | 2 - 3 | 230 | 40106771 | 77 |
BC118046.1 | - | 2 - 3 | 230 | 40106772 | 77 | |
U29508.1 | - | 2 - 3 | 145 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |