Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10599287 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599290 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599250 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599289 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599247 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599288 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599249 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10599248 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1420448_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10599286 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10599246 |
- |
- |
Mouse Genome 430 2.0 Array |
1420448_at |
- |
- |
Mouse Genome 430 2.0 Array |
1420448_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00002302.mm.1 |
NM_001099316.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002302.mm.1 |
NM_001099316.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002302.mm.1 |
TC0X00000343.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002354.mm.1 |
NM_001099318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002354.mm.1 |
NM_001099318.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002354.mm.1 |
TC0X00003391.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002245.mm.1 |
NM_029203.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002245.mm.1 |
NM_029203.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002245.mm.1 |
TC0X00000338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002299.mm.1 |
NM_001099316.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002299.mm.1 |
NM_001099316.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002299.mm.1 |
TC0X00000343.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002300.mm.1 |
NM_001099316.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002300.mm.1 |
NM_001099316.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002300.mm.1 |
TC0X00000343.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002301.mm.1 |
NM_001099316.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002301.mm.1 |
NM_001099316.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002301.mm.1 |
TC0X00000343.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002401.mm.1 |
NM_001085348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002401.mm.1 |
NM_001085348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002401.mm.1 |
TC0X00000350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002243.mm.1 |
NM_029203.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002243.mm.1 |
NM_029203.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002243.mm.1 |
TC0X00000338.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00002246.mm.1 |
NM_029203.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002246.mm.1 |
NM_029203.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00002246.mm.1 |
TC0X00000338.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1420448_at |
- |
- |
Mouse Expression Set 430 |
1420448_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420448_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420448_PM_at |
- |
- |