Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102457.4 | NP_001095927.3 | 1 - 2 | 180 | 100 | |
GenBank mRNA | BC087875.1 | - | 1 - 2 | 51 | 100 | |
BC092527.1 | - | 1 - 2 | 51 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |