Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 34 - 35 | 7377 | 82 | |
NM_010862.2 | NP_034992.2 | 35 - 36 | 7431 | 82 | ||
NM_182698.2 | NP_874357.2 | 33 - 34 | 3615 | 82 | ||
GenBank mRNA | AF053130.1 | - | 32 - 33 | 3583 | 82 | |
AF144095.1 | - | 35 - 36 | 7431 | 82 | ||
AY331132.1 | - | 34 - 35 | 7377 | 82 | ||
AY331133.1 | - | 33 - 34 | 3615 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |