Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 45 - 46 | 8518 | 92 | |
NM_010862.2 | NP_034992.2 | 46 - 47 | 8572 | 92 | ||
NM_182698.2 | NP_874357.2 | 44 - 45 | 4756 | 92 | ||
GenBank mRNA | AF053130.1 | - | 43 - 44 | 4724 | 92 | |
AF144095.1 | - | 46 - 47 | 8572 | 92 | ||
AY331132.1 | - | 45 - 46 | 8518 | 92 | ||
AY331133.1 | - | 44 - 45 | 4756 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |