Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 6 - 7 | 4184 | 99 | |
NM_010862.2 | NP_034992.2 | 6 - 7 | 4184 | 99 | ||
NM_182698.2 | NP_874357.2 | 5 - 6 | 422 | 99 | ||
GenBank mRNA | AB014510.1 | - | 2 - 3 | 187 | 99 | |
AF053130.1 | - | 5 - 6 | 396 | 99 | ||
AF144095.1 | - | 6 - 7 | 4184 | 99 | ||
AY331132.1 | - | 6 - 7 | 4184 | 99 | ||
AY331133.1 | - | 5 - 6 | 422 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |