Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700020326.mm.1 |
TC0700002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020326.mm.1 |
NM_001105068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020326.mm.1 |
NM_001105068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020518.mm.1 |
TC0700002275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020518.mm.1 |
NM_001105072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020518.mm.1 |
NM_001105072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020672.mm.1 |
TC0700002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020672.mm.1 |
NM_001105152.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020672.mm.1 |
NM_001105152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020325.mm.1 |
TC0700002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020325.mm.1 |
NM_001105068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020325.mm.1 |
NM_001105068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020380.mm.1 |
TC0700002253.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020380.mm.1 |
NM_001105073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020380.mm.1 |
NM_001105073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020519.mm.1 |
TC0700002275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020519.mm.1 |
NM_001105072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020519.mm.1 |
NM_001105072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020324.mm.1 |
TC0700002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020324.mm.1 |
NM_001105068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020324.mm.1 |
NM_001105068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020381.mm.1 |
TC0700002253.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020381.mm.1 |
NM_001105073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020381.mm.1 |
NM_001105073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020673.mm.1 |
TC0700002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020673.mm.1 |
NM_001105152.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020673.mm.1 |
NM_001105152.1 |
Refseq |