Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Perp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022032.4 | NP_071315.1 | 1 - 2 | 334 | 103 | |
GenBank mRNA | AF249870.1 | - | 1 - 2 | 335 | 103 | |
AK003734.1 | - | 1 - 2 | 275 | 103 | ||
AK009184.1 | - | 1 - 2 | 272 | 103 | ||
AK009190.1 | - | 1 - 2 | 273 | 103 | ||
BC021772.1 | - | 1 - 2 | 259 | 3985702 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |