Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159750.1 | NP_001153222.1 | 5 - 6 | 684 | 94 | |
NM_001159751.1 | NP_001153223.1 | 5 - 6 | 580 | 94 | ||
NM_011541.4 | NP_035671.1 | 5 - 6 | 687 | 94 | ||
GenBank mRNA | AK131928.1 | - | 6 - 7 | 871 | 94 | |
AK136035.1 | - | 5 - 6 | 580 | 94 | ||
AK143593.1 | - | 5 - 6 | 545 | 94 | ||
BC006022.1 | - | 5 - 6 | 562 | 3590733 | 94 | |
BC061490.1 | - | 5 - 6 | 579 | 2582185 | 94 | |
BC083127.1 | - | 5 - 6 | 550 | 6390364 | 94 | |
M18209.1 | - | 4 - 5 | 483 | 94 | ||
M18210.1 | - | 5 - 6 | 541 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |