Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001024619.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001024619.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001168539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001168539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001168540.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001168540.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001168541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
NM_001168541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031854.mm.1 |
TC0700003778.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031861.mm.1 |
NM_001168539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031861.mm.1 |
NM_001168539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031861.mm.1 |
TC0700003778.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001024619.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001024619.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001168539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001168539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001168540.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001168540.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001168541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
NM_001168541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031855.mm.1 |
TC0700003778.mm.1 |
MTA TC |