Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009025.2 | NP_033051.2 | 7 - 8 | 741 | 94 | |
GenBank mRNA | AK053353.1 | - | 7 - 8 | 737 | 94 | |
AK141511.1 | - | 7 - 8 | 748 | 94 | ||
AK147896.1 | - | 7 - 8 | 709 | 94 | ||
BC057300.1 | - | 7 - 8 | 732 | 6821254 | 94 | |
BC060195.1 | - | 7 - 8 | 756 | 94 | ||
BC068297.1 | - | 7 - 8 | 605 | 30102545 | 94 | |
U20238.1 | - | 7 - 8 | 726 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |