Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009136.3 | NP_033162.1 | 1 - 2 | 209 | 104 | |
GenBank mRNA | AJ223206.1 | - | 1 - 2 | 209 | 104 | |
AK046213.1 | - | 1 - 2 | 78 | 104 | ||
BC058532.1 | - | 97 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |