Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009136.3 | NP_033162.1 | 2 - 3 | 456 | 88 | |
GenBank mRNA | AJ223206.1 | - | 2 - 3 | 456 | 88 | |
AK046213.1 | - | 2 - 3 | 325 | 88 | ||
BC058532.1 | - | 344 | 88 | |||
CT010374.1 | - | 1 - 2 | 240 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |