Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200015827.mm.1 |
NM_009244.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015827.mm.1 |
NM_009244.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015827.mm.1 |
TC1200002531.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015866.mm.1 |
NM_009245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015866.mm.1 |
NM_009245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015866.mm.1 |
TC1200002534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015826.mm.1 |
NM_009244.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015826.mm.1 |
NM_009244.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015826.mm.1 |
TC1200002531.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200018825.mm.1 |
NM_001252569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018825.mm.1 |
NM_001252569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018825.mm.1 |
NM_009243.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018825.mm.1 |
NM_009243.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018825.mm.1 |
TC1200002533.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015875.mm.1 |
NM_009245.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015875.mm.1 |
NM_009245.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015875.mm.1 |
TC1200002534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200018822.mm.1 |
NM_009244.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018822.mm.1 |
NM_009244.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018822.mm.1 |
TC1200002531.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200018824.mm.1 |
NM_001252569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018824.mm.1 |
NM_001252569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018824.mm.1 |
NM_009243.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018824.mm.1 |
NM_009243.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200018824.mm.1 |
TC1200002533.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015861.mm.1 |
NM_001252569.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015861.mm.1 |
NM_001252569.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015861.mm.1 |
NM_009243.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015861.mm.1 |
NM_009243.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015861.mm.1 |
TC1200002533.mm.1 |
MTA TC |