Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019718.mm.1 |
NM_009638.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019718.mm.1 |
NM_009638.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019718.mm.1 |
TC1700002103.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019716.mm.1 |
NM_009638.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019716.mm.1 |
NM_009638.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019716.mm.1 |
TC1700002103.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019717.mm.1 |
NM_009638.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019717.mm.1 |
NM_009638.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019717.mm.1 |
TC1700002103.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019714.mm.1 |
NM_009638.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019714.mm.1 |
NM_009638.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019714.mm.1 |
TC1700002103.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019715.mm.1 |
NM_009638.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019715.mm.1 |
NM_009638.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019715.mm.1 |
TC1700002103.mm.1 |
MTA TC |