Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010380.3 | NP_034510.3 | 3 - 4 | 641 | 121 | |
GenBank mRNA | AK159572.1 | - | 3 - 4 | 642 | 121 | |
BC023409.1 | - | 2 - 3 | 533 | 121 | ||
BC039637.1 | - | 2 - 3 | 337 | 121 | ||
BC092284.1 | - | 438 | 121 | |||
BC106173.1 | - | 566 | 121 | |||
BC132088.1 | - | 3 - 4 | 709 | 40130365 | 121 | |
BC132655.1 | - | 3 - 4 | 709 | 40130932 | 121 | |
K00129.1 | - | 2 - 3 | 308 | 121 | ||
KF831063.1 | - | 3 - 4 | 612 | 121 | ||
KF831067.1 | - | 3 - 4 | 612 | 121 | ||
L36068.1 | - | 3 - 4 | 621 | 121 | ||
M12185.1 | - | 2 - 3 | 308 | 121 | ||
M37680.1 | - | 2 - 3 | 548 | 121 | ||
M37681.1 | - | 2 - 3 | 548 | 121 | ||
M37682.1 | - | 2 - 3 | 548 | 121 | ||
U47325.1 | - | 3 - 4 | 621 | 121 | ||
X52490.1 | - | 3 - 4 | 630 | 121 | ||
Y00806.1 | - | 2 - 3 | 548 | 121 |
Gene Symbol: H2-K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001892.2 | NP_001001892.2 | 3 - 4 | 121 | ||
GenBank mRNA | AF470231.1 | - | 3 - 4 | 121 | ||
AK150790.1 | - | 3 - 4 | 121 | |||
AK150967.1 | - | 3 - 4 | 121 | |||
AK153419.1 | - | 3 - 4 | 121 | |||
AK159321.1 | - | 3 - 4 | 121 | |||
AK160102.1 | - | 3 - 4 | 121 | |||
AK168517.1 | - | 3 - 4 | 121 | |||
BC151193.1 | - | 3 - 4 | 9088081 | 121 | ||
J00400.1 | - | 2 - 3 | 121 | |||
JX316765.1 | - | 121 | ||||
KF831064.1 | - | 3 - 4 | 121 | |||
KF831065.1 | - | 3 - 4 | 121 | |||
KF831066.1 | - | 3 - 4 | 121 | |||
L23494.1 | - | 3 - 4 | 121 | |||
L23495.1 | - | 3 - 4 | 121 | |||
M11975.1 | - | 3 - 4 | 121 | |||
M34932.1 | - | 121 | ||||
M58156.1 | - | 3 - 4 | 121 | |||
M60168.1 | - | 3 - 4 | 121 | |||
M60169.1 | - | 3 - 4 | 121 | |||
M69070.1 | - | 3 - 4 | 121 | |||
M69071.1 | - | 3 - 4 | 121 | |||
M69072.1 | - | 3 - 4 | 121 | |||
U47328.1 | - | 3 - 4 | 121 | |||
U47330.1 | - | 3 - 4 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |