Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010734.2 | NP_034864.2 | 2 - 3 | 180 | 79 | |
GenBank mRNA | AI182980.1 | - | 2 - 3 | 180 | 79 | |
AI323110.1 | - | 1 - 2 | 85 | 79 | ||
AI482475.1 | - | 2 - 3 | 79 | |||
BC086881.1 | - | 2 - 3 | 174 | 6747449 | 79 | |
M18187.1 | - | 2 - 3 | 173 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |