Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100008753.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008753.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008753.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008756.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008756.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008756.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008751.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008751.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008751.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008752.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008752.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008752.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008747.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008747.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008747.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008754.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008754.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008754.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008748.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008748.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008748.mm.1 |
TC1100000975.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008750.mm.1 |
NM_025993.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008750.mm.1 |
NM_025993.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008750.mm.1 |
TC1100000975.mm.1 |
MTA TC |