Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026424.3 | NP_080700.3 | 1 - 2 | 222 | 100 | |
GenBank mRNA | AK006551.1 | - | 1 - 2 | 223 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |