Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027191.2 | NP_081467.2 | 4 - 5 | 499 | 120 | |
NM_028334.4 | NP_082610.1 | 4 - 5 | 517 | 120 | ||
GenBank mRNA | AK010370.1 | - | 4 - 5 | 443 | 120 | |
AK012890.1 | - | 4 - 5 | 120 | |||
AK040463.1 | - | 4 - 5 | 500 | 120 | ||
AK165537.1 | - | 4 - 5 | 463 | 120 | ||
BC011102.1 | - | 4 - 5 | 483 | 3982076 | 120 | |
BC032180.1 | - | 3 - 4 | 427 | 5151847 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |