Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029044.2 | NP_083320.1 | 5 - 6 | 627 | 93 | |
GenBank mRNA | AK015430.1 | - | 5 - 6 | 629 | 93 | |
AK047468.1 | - | 5 - 6 | 682 | 92 | ||
AK049695.1 | - | 5 - 6 | 655 | 93 | ||
BC018412.1 | - | 5 - 6 | 640 | 3673669 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |