Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139139.2 | NP_631878.2 | 9 - 10 | 714 | 97 | |
GenBank mRNA | AK077385.1 | - | 9 - 10 | 674 | 97 | |
BC013487.1 | - | 9 - 10 | 693 | 4211118 | 97 | |
BP770408.1 | - | 2 - 3 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |