Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013555.4 | NP_038583.1 | 1 - 2 | 1070 | 53 | |
GenBank mRNA | AK134427.1 | - | 1 - 2 | 860 | 53 | |
AK144746.1 | - | 1 - 2 | 215 | 53 | ||
BC019150.1 | - | 1 - 2 | 1105 | 5064578 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |