Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013621.3 | NP_038649.2 | 1 - 2 | 51 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons |