Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013717.2 | NP_038745.1 | 3 - 4 | 281 | 108 | |
GenBank mRNA | AB030483.1 | - | 3 - 4 | 281 | 108 | |
AK003038.1 | - | 3 - 4 | 276 | 108 | ||
AK010355.1 | - | 3 - 4 | 310 | 108 | ||
AK131701.1 | - | 3 - 4 | 272 | 108 | ||
BC008113.1 | - | 3 - 4 | 249 | 3483517 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |