Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016668.3 | NP_057877.1 | 5 - 6 | 886 | 67 | |
GenBank mRNA | AF033381.1 | - | 5 - 6 | 629 | 67 | |
AK145668.1 | - | 5 - 6 | 728 | 67 | ||
AK149457.1 | - | 5 - 6 | 693 | 67 | ||
AK166754.1 | - | 5 - 6 | 695 | 67 | ||
AK166818.1 | - | 5 - 6 | 694 | 67 | ||
BC037004.1 | - | 5 - 6 | 688 | 5100429 | 67 | |
BC093510.1 | - | 5 - 6 | 661 | 5101184 | 67 | |
BC110307.1 | - | 5 - 6 | 1089 | 5310035 | 67 | |
BG078929.2 | - | 3 - 4 | 378 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |