Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173747.3 | NP_776108.2 | 3 - 4 | 545 | 101 | |
GenBank mRNA | AK028433.1 | - | 3 - 4 | 467 | 101 | |
AK144167.1 | - | 3 - 4 | 475 | 101 | ||
BC062646.1 | - | 1 - 2 | 95 | 101 | ||
BC092224.1 | - | 3 - 4 | 451 | 30355916 | 101 | |
BC120679.1 | - | 2 - 3 | 228 | 40129602 | 101 | |
BC125335.1 | - | 2 - 3 | 228 | 40129850 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |