Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stfa2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173869.3 | NP_776294.1 | 1 - 2 | 113 | 106 | |
GenBank mRNA | AK050597.1 | - | 1 - 2 | 114 | 106 | |
AY163161.1 | - | 1 - 2 | 89 | 106 | ||
BC117052.1 | - | 1 - 2 | 111 | 40126371 | 106 | |
BC119354.1 | - | 1 - 2 | 111 | 8734103 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |