Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177348.2 | NP_796322.1 | 1 | 247 | 88 | |
GenBank mRNA | AY190044.1 | - | 1 | 247 | 88 | |
BC104376.2 | - | 1 | 246 | 40050053 | 88 | |
BC104377.2 | - | 1 | 246 | 40050054 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |