Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177850.3 | NP_808518.1 | 10 - 11 | 1218 | 102 | |
GenBank mRNA | AK033770.1 | - | 10 - 11 | 1220 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |