Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252519.1 | NP_001239448.1 | 2 - 3 | 160 | 86 | |
NM_001253385.1 | NP_001240314.1 | 2 - 3 | 160 | 86 | ||
NM_177889.5 | NP_808557.2 | 2 - 3 | 160 | 86 | ||
GenBank mRNA | AK037211.1 | - | 2 - 3 | 157 | 86 | |
BC060530.1 | - | 1 - 2 | 87 | 5684819 | 86 | |
BY127198.1 | - | 2 - 3 | 160 | 86 | ||
CD806198.1 | - | 1 - 2 | 111 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |