Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ispd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289502.1 | NP_001276431.1 | 7 - 8 | 1283 | 94 | |
NM_001289503.1 | NP_001276432.1 | 8 - 9 | 1015 | 94 | ||
NM_178629.6 | NP_848744.2 | 8 - 9 | 1433 | 94 | ||
GenBank mRNA | AK016317.1 | - | 8 - 9 | 981 | 94 | |
AK045940.1 | - | 8 - 9 | 1435 | 94 | ||
AK051322.1 | - | 7 - 8 | 978 | 94 | ||
BC024061.1 | - | 7 - 8 | 715 | 94 | ||
BC050993.1 | - | 8 - 9 | 1271 | 94 | ||
BC086665.1 | - | 8 - 9 | 1364 | 30612014 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |