Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700024270.mm.1 |
TC0700002740.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024270.mm.1 |
NM_011748.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024270.mm.1 |
NM_011748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024270.mm.1 |
NM_178733.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024270.mm.1 |
NM_178733.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024272.mm.1 |
TC0700002740.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024272.mm.1 |
NM_011748.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024272.mm.1 |
NM_011748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024272.mm.1 |
NM_178733.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024272.mm.1 |
NM_178733.5 |
Refseq |