Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssxb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199066.2 | NP_951021.2 | 4 - 5 | 458 | 83 | |
GenBank mRNA | AY244800.1 | - | 458 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |