Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201355.3 | NP_958743.1 | 1 - 2 | 393 | 103 | |
GenBank mRNA | AK158627.1 | - | 1 - 2 | 394 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |