Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700020099.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020099.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020099.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020094.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020094.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020094.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020105.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020105.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020105.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020103.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020103.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020103.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020100.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020100.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020100.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020090.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020090.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020090.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020096.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020096.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020096.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020095.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020095.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020095.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020097.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020097.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020097.mm.1 |
TC1700002151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700020102.mm.1 |
NM_008302.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020102.mm.1 |
NM_008302.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700020102.mm.1 |
TC1700002151.mm.1 |
MTA TC |