Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011321.3 | NP_035451.1 | 1 | 47 | 75 | |
GenBank mRNA | AK035423.1 | - | 1 | 48 | 75 | |
BC049605.1 | - | 1 | 47 | 6743567 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |