Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145534.2 | NP_663509.2 | 1 | 450 | 90 | |
GenBank mRNA | AB093275.1 | - | 1 | 450 | 90 | |
BC018174.1 | - | 1 | 421 | 4011475 | 90 | |
BC062968.1 | - | 1 | 146 | 6408511 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |