Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twistnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172253.2 | NP_758457.1 | 1 | 46 | 105 | |
GenBank mRNA | AK010829.1 | - | 1 | 47 | 105 | |
AK016638.1 | - | 1 | 43 | 105 | ||
AK017016.1 | - | 1 | 36 | 105 | ||
AK153090.1 | - | 36 | 105 | |||
AK160682.1 | - | 1 | 34 | 105 | ||
BC139162.1 | - | 1 | 34 | 8862184 | 105 | |
BC139163.1 | - | 1 | 34 | 8862185 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |