Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ND4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140187.1 | - | 1 | 54 | 85 | |
AK144853.1 | - | 54 | 85 | |||
AK153747.1 | - | 53 | 85 | |||
AK157367.1 | - | 54 | 85 | |||
NC_005089_ND4L.0 | - | 53 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mitochondria Encoded Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon |