Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027953.1 | 1 - 2 | 130 | 115 | ||
GenBank mRNA | AA268702.1 | - | 1 - 2 | 84 | 115 | |
AI265689.1 | - | 2 - 3 | 115 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | speedy homolog A (Xenopus laevis) | Spdya |
4921517J08Rik, 4930548B21Rik, GS4, MLZ-465, Spdy1 |
-953.0 | Anti-sense | Chr. 17 - 71526937 - 71589535 on NCBI Build GRCm38 |
Downstream | WD repeat domain 43 | Wdr43 |
2610318G08Rik, AU020887, AV024208, Wrd43, mKIAA0007 |
17.6kb | Anti-sense | Chr. 17 - 71615900 - 71659031 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |