Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900016814.mm.1 |
NM_001199953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016814.mm.1 |
NM_001199953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016814.mm.1 |
TC0900002023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016816.mm.1 |
NM_001199953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016816.mm.1 |
NM_001199953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016816.mm.1 |
TC0900002023.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900016817.mm.1 |
NM_001199953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016817.mm.1 |
NM_001199953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900016817.mm.1 |
TC0900002023.mm.1 |
MTA TC |