Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080729.3 | NP_542767.1 | 1 - 2 | 89 | 57 | |
GenBank mRNA | BC120568.1 | - | 1 - 2 | 88 | 40129491 | 57 |
BC125524.1 | - | 1 - 2 | 88 | 40130039 | 57 | |
BC144962.1 | - | 1 - 2 | 88 | 9053510 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |