Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290740.1 | NP_001277669.1 | 6 - 7 | 891 | 71 | |
NM_030560.5 | NP_085037.2 | 7 - 8 | 949 | 71 | ||
NM_172667.2 | NP_766255.1 | 6 - 7 | 888 | 71 | ||
GenBank mRNA | AK045589.1 | - | 6 - 7 | 720 | 71 | |
AK077961.1 | - | 7 - 8 | 777 | 71 | ||
AK089988.1 | - | 727 | 71 | |||
AK129404.1 | - | 705 | 71 | |||
AK132001.1 | - | 6 - 7 | 832 | 71 | ||
AK143352.1 | - | 6 - 7 | 721 | 71 | ||
AK149528.1 | - | 6 - 7 | 715 | 71 | ||
AK166487.1 | - | 6 - 7 | 727 | 71 | ||
BC003993.1 | - | 6 - 7 | 701 | 3492146 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |