Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182808.3 | NP_877960.1 | 2 - 3 | 555 | 63 | |
GenBank mRNA | AK049880.1 | - | 1 - 2 | 446 | 63 | |
AK050001.1 | - | 2 - 3 | 556 | 63 | ||
AY325120.1 | - | 2 - 3 | 356 | 63 | ||
BC132179.1 | - | 2 - 3 | 390 | 40130456 | 63 | |
BC132181.1 | - | 2 - 3 | 390 | 40130458 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |