Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281926.1 | NP_001268855.1 | 1 - 2 | 127 | 91 | |
GenBank mRNA | AK147620.1 | - | 1 - 2 | 128 | 91 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | eyes absent 2 homolog (Drosophila) | Eya2 | Eab1 | -104.0kb | Anti-sense | Chr. 2 - 165595028 - 165771727 on NCBI Build GRCm38 |
Downstream | nuclear receptor coactivator 3 | Ncoa3 |
2010305B15Rik, AW321064, Actr, Aib1, KAT13B, Rac3, Src3, Tram-1, Tram1, bHLHe42, p/Cip, pCip |
110.4kb | Anti-sense | Chr. 2 - 165992637 - 166073242 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |